missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum Chloride Releasing Agent: 1% La (from oxide) in 2% HCl, VHG™

Product Code. 30003112
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Product Code. Quantity unitSize
30003112 500 mL 500mL
30002947 100 mL 100mL
2 options
This item is not returnable. View return policy

Product Code. 30003112

Brand: VHG Labs VHGMLAH1P500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Lanthanum Chloride Releasing Agent: 1% La (from oxide) in 2% HCl
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lanthanum
CAS 7439-91-0
Methods AA/GFAA
Analyte or Component Names Lanthanum
Shipping Condition Room Temperature
Molecular Formula La
Synonym La
InChI Formula InChI=1S/La
SMILES [La]
IUPAC Name lanthanum
Solvent or Matrix Dilute HCl
Molecular Weight (g/mol) 138.906
Quantity 500 mL
Formula Weight 138.906 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.