missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum aluminum oxide substrate, 10x10x1mm, polished one side, 100 orientation

Product Code. p-4905390
Change view
Click to view available options
Quantity:
1 Ea.
5 Ea.
Unit Size:
Each
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11358837 1 Ea. Each
11368837 5 Ea. Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11358837 Supplier Thermo Scientific Alfa Aesar Supplier No. 038495.KT
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

100 Orientation | CAS: 12003-65-5 | LaAlO3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12003-65-5
Molecular Formula LaAlO3
MDL Number MFCD00144550

Specifications

Color Brown-Yellow
Physical Form Plate
Quantity 1 Ea.
Odor Odorless
Chemical Name or Material Lanthanum Aluminum Oxide Substrate, Polished Two Sides

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.