missing translation for 'onlineSavingsMsg'
Learn More

Lansoprazole, TRC

Product Code. 30133851
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133851 1 g 1g
30134153 250 mg 250mg
30003384 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133851 Supplier Toronto Research Chemicals Supplier No. TRCL1750001G
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103577-45-3
Molecular Formula C16 H14 F3 N3 O2 S
Molecular Weight (g/mol) 369.36
Synonym Lansoprazole, (±)-Lansoprazole, 2-[[[3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridyl]methyl]sulfinyl]-1H-benzimidazole, A 65006, AG 1749, Agopton, Ilsatec, Ketian, Lancid, Lanfast, Lanproton, Lansopep, Lansophed, Lanston, Lanz, Lanzol 30, Lanzopral, Lanzor, Lapraz, Ogast, Ogastro, PP/K-10, Prevacid, Promp, Prosogan, Suprecid, Takepron, Ulpax, Zoton
IUPAC Name 2-[[3-methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl]methylsulfinyl]-1H-benzimidazole
SMILES Cc1c(OCC(F)(F)F)ccnc1CS(=O)c2nc3ccccc3[nH]2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C16H14F3N3O2S/c1-10-13(20-7-6-14(10)24-9-16(17,18)19)8-25(23)15-21-11-4-2-3-5-12(11)22-15/h2-7H,8-9H2,1H3,(H,21,22)
Formula Weight 369.0759
Chemical Name or Material Lansoprazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.