missing translation for 'onlineSavingsMsg'
Learn More

Lansoprazole Sulphone N-Oxide, Mikromol™

Product Code. 30051134
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051134 25 mg 25mg
30051072 100 mg 100mg
30459930 4 x 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30051134 Supplier Mikromol Supplier No. MM0474.080025
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Lansoprazole Sulphone N-Oxide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lansoprazole Sulphone N-Oxide
CAS 953787-54-7
Analyte or Component Names Lansoprazole Sulphone N-Oxide
Shipping Condition Room Temperature
Molecular Formula C16 H14 F3 N3 O4 S
Certifications/Compliance ISO 9001
Synonym Lansoprazole Sulphone N-Oxide, Lansoprazole Sulfone N-Oxide
InChI Formula InChI=1S/C16H14F3N3O4S/c1-10-13(22(23)7-6-14(10)26-9-16(17,18)19)8-27(24,25)15-20-11-4-2-3-5-12(11)21-15/h2-7H,8-9H2,1H3,(H,20,21)
SMILES Cc1c(OCC(F)(F)F)cc[n+]([O-])c1CS(=O)(=O)c2nc3ccccc3[nH]2
IUPAC Name 2-[[3-methyl-1-oxido-4-(2,2,2-trifluoroethoxy)pyridin-1-ium-2-yl]methylsulfonyl]-1H-benzimidazole
Molecular Weight (g/mol) 401.36
Quantity 25 mg
Formula Weight 401.0657 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.