missing translation for 'onlineSavingsMsg'
Learn More

Lansoprazole Sulfone N-Oxide, TRC

Product Code. 30138774
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138774 25 mg 25mg
30134249 50 mg 50mg
30011680 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138774 Supplier Toronto Research Chemicals Supplier No. TRCL17503025MG
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 953787-54-7
Molecular Formula C16 H14 F3 N3 O4 S
Molecular Weight (g/mol) 401.36
Synonym Lansoprazole Sulphone N-Oxide, Lansoprazole Sulfone N-Oxide
IUPAC Name 2-[[3-methyl-1-oxido-4-(2,2,2-trifluoroethoxy)pyridin-1-ium-2-yl]methylsulfonyl]-1H-benzimidazole
SMILES Cc1c(OCC(F)(F)F)cc[n+]([O-])c1CS(=O)(=O)c2nc3ccccc3[nH]2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C16H14F3N3O4S/c1-10-13(22(23)7-6-14(10)26-9-16(17,18)19)8-27(24,25)15-20-11-4-2-3-5-12(11)21-15/h2-7H,8-9H2,1H3,(H,20,21)
Formula Weight 401.0657
Chemical Name or Material Lansoprazole Sulphone N-Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.