missing translation for 'onlineSavingsMsg'
Learn More

Lansoprazole, 98+%

Product Code. 15494469
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15494469 1 g 1g
15414479 500 mg 500mg
15404479 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15494469 Supplier Thermo Scientific Alfa Aesar Supplier No. J62008.03
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A proton pump inhibitor | CAS: 103577-45-3 | C16H14F3N3O2S | 369.362 g/mol

A proton pump inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103577-45-3
Molecular Formula C16H14F3N3O2S
Molecular Weight (g/mol) 369.362
MDL Number MFCD00866873
InChI Key MJIHNNLFOKEZEW-UHFFFAOYSA-N
Synonym lansoprazole, prevacid, bamalite, monolitum, lansoprazol, agopton, limpidex, ogastro, lanzor, opiren
PubChem CID 3883
ChEBI CHEBI:6375
IUPAC Name 2-[[3-methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl]methylsulfinyl]-1H-benzimidazole
SMILES CC1=C(C=CN=C1CS(=O)C2=NC3=CC=CC=C3N2)OCC(F)(F)F

Specifications

Melting Point 173°C to 175°C
Color White
Quantity 1 g
Merck Index 14,5362
Formula Weight 369.36
Percent Purity ≥98%
Physical Form Crystalline Powder
Chemical Name or Material Lansoprazole

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.