missing translation for 'onlineSavingsMsg'
Learn More

Lansoprazole, 98+%, Thermo Scientific™

A proton pump inhibitor

Brand:  Thermo Scientific Alfa Aesar J62008.06

Quantity 5g

 View more versions of this product

Product Code. 15404479

  • £547.00 / 5g
 Restricted Item
Estimated Shipment: 17-09-2026
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

103577-45-3
369.362
MJIHNNLFOKEZEW-UHFFFAOYSA-N
3883
2-[[3-methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl]methylsulfinyl]-1H-benzimidazole
C16H14F3N3O2S
MFCD00866873
lansoprazole, prevacid, bamalite, monolitum, lansoprazol, agopton, limpidex, ogastro, lanzor, opiren
CHEBI:6375
CC1=C(C=CN=C1CS(=O)C2=NC3=CC=CC=C3N2)OCC(F)(F)F

Specifications

103577-45-3
MFCD00866873
14,5362
MJIHNNLFOKEZEW-UHFFFAOYSA-N
2-[[3-methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl]methylsulfinyl]-1H-benzimidazole
3883
369.36
Lansoprazole
C16H14F3N3O2S
5g
lansoprazole, prevacid, bamalite, monolitum, lansoprazol, agopton, limpidex, ogastro, lanzor, opiren
CC1=C(C=CN=C1CS(=O)C2=NC3=CC=CC=C3N2)OCC(F)(F)F
369.362
CHEBI:6375
≥98%
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.