missing translation for 'onlineSavingsMsg'
Learn More

Lanosterol, MedChemExpress

Product Code. 16443459
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16443459 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16443459 Supplier Medchem Express Supplier No. HYW020033
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Lanosterol is an intermediate of cholesterol synthesis and use of lanosterol induces ubiquitination and degradation of a rate-controlling enzyme of cholesterol synthesis, i.e., HMG CoA reductase. Lanosterol suppresses the aggregation and cytotoxicity of misfolded proteins linked with neurodegenerative diseases.

Lanosterol is an intermediate of cholesterol synthesis and use of lanosterol induces ubiquitination and degradation of a rate-controlling enzyme of cholesterol synthesis, i.e., HMG CoA reductase. Lanosterol suppresses the aggregation and cytotoxicity of misfolded proteins linked with neurodegenerative diseases.

Specifications

Chemical Name or Material Lanosterol
CAS 79-63-0
Color White
Purity Grade Notes Research
Molecular Formula C30H50O
Quantity 1 mg
Solubility Information Ethanol : 10 mg/mL (23.43 mM; ultrasonic and warming and heat to 60°C) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble)
SMILES C[C@]12C3=C(CC[C@@]1([C@]([C@H](C)CC/C=C(C)/C)([H])CC2)C)[C@@]4([C@@](C(C)([C@@H](O)CC4)C)([H])CC3)C
Molecular Weight (g/mol) 426.72
Formula Weight 426.72
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.