missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lanopepden, MedChemExpress
Click to view available options
Quantity:
1 mg
2 mg
5 mg
Unit Size:
1mg
2mg
5mg
Description
Lanopepden (GSK 1322322) is a peptide deformylase inhibitor active against Staphylococcus aureus strains with MICs of 1 and 1 mg/L for ATCC 29213 and ATCC 25923 strain, respectively.
Specifications
Specifications
| Chemical Name or Material | Lanopepden |
| Physical Form | Solid |
| CAS | 1152107-25-9 |
| Color | Light Yellow |
| Quantity | 1 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C22H34FN7O4 |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | GSK 1322322 |
| SMILES | FC1=C(N2CCN(CCOC3)[C@@]3([H])C2)N=C(C)N=C1NNC([C@H](CC4CCCC4)CN(C=O)O)=O |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction