missing translation for 'onlineSavingsMsg'
Learn More

Langerin/CD207 Antibody (LGRN/7541), Novus Biologicals™

Mouse Monoclonal Antibody

£195.00 - £385.00

Specifications

Antigen Langerin/CD207
Clone LGRN/7541
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 μg/mL
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18620005
View Documents
Novus Biologicals
NBP3-20557-20ug
20 μg
£195.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18623545
View Documents
Novus Biologicals
NBP3-20557-100ug
100 μg
£385.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Langerin/CD207 Monoclonal antibody specifically detects Langerin/CD207 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

Langerin/CD207
0.2 mg/ml
Immunohistochemistry (Paraffin)
Unconjugated
Mouse
metabolism, Signal Transduction
10mM PBS with 0.05% BSA
50489
IgG
Protein A or G purified
LGRN/7541
Immunohistochemistry-Paraffin 1-2 μg/mL
Monoclonal
Purified
RUO
Human
CD207 antigen, CD207 antigen, langerin, CD207 molecule, langerin, CLEC4KLangerhans cell specific c-type lectin, C-type lectin domain family 4 member K, C-type lectin domain family 4, member K, Langerin
Recombinant fragment (around aa50-250) of human Langerin/CD207 protein (exact sequence is proprietary)
Primary
Store at 4&drg;C. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.