missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Langerin/CD207 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
£181.00 - £433.00
Specifications
| Antigen | Langerin/CD207 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1 - 3 ug/ml, Flow Cytometry 1:20-1:2000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, Immunohistochemistry-Frozen 1:200-1:1000, Flow (Intracellular) reported in scientific literature (PMID 18369922) |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18779123
|
Novus Biologicals
NB100-56733SS |
0.025 mg |
£181.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18728883
|
Novus Biologicals
NB100-56733 |
0.1 mg |
£433.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Langerin/CD207 Polyclonal specifically detects Langerin/CD207 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular).Specifications
| Langerin/CD207 | |
| Western Blot 1 - 3 ug/ml, Flow Cytometry 1:20-1:2000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, Immunohistochemistry-Frozen 1:200-1:1000, Flow (Intracellular) reported in scientific literature (PMID 18369922) | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| Q8VBX4 | |
| 50489 | |
| Amino acids 26-44 of mouse Langerin were used as immunogen (Uniprot # Q8VBX4) | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Rabbit | |
| metabolism, Signal Transduction | |
| PBS with 0.05% Sodium Azide | |
| CD207 antigen, CD207 antigen, langerin, CD207 molecule, langerin, CLEC4KLangerhans cell specific c-type lectin, C-type lectin domain family 4 member K, C-type lectin domain family 4, member K, Langerin | |
| CD207 | |
| IgG | |
| Protein G purified | |
| Cross reacts with Mouse. Predicted to react with Rat. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title