missing translation for 'onlineSavingsMsg'
Learn More

Landiolol Hydrochloride, TRC

Product Code. 30007336
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007336 100 mg 100mg
30005657 10 mg 10mg
30011890 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007336 Supplier Toronto Research Chemicals Supplier No. TRCL173900100MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144481-98-1
Molecular Formula C25 H39 N3 O8 . Cl H
Molecular Weight (g/mol) 546.05
Synonym Benzenepropanoic acid, 4-[(2S)-2-hydroxy-3-[[2-[(4-morpholinylcarbonyl)amino]ethyl]amino]propoxy]-, [(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]methyl ester, hydrochloride (1:1), Benzenepropanoic acid, 4-[(2S)-2-hydroxy-3-[[2-[(4-morpholinylcarbonyl)amino]ethyl]amino]propoxy]-, [(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]methyl ester, monohydrochloride (9CI), Benzenepropanoic acid, 4-[2-hydroxy-3-[[2-[(4-morpholinylcarbonyl)amino]ethyl]amino]propoxy]-, (2,2-dimethyl-1,3-dioxolan-4-yl)methyl ester, monohydrochloride, [S-(R*,R*)]-, Corebeta, Landiolol hydrochloride, ONO 1101 hydrochloride, Onoact
IUPAC Name [(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]methyl 3-[4-[(2S)-2-hydroxy-3-[2-(morpholine-4-carbonylamino)ethylamino]propoxy]phenyl]propanoate;hydrochloride
SMILES Cl.CC1(C)OC[C@@H](COC(=O)CCc2ccc(OC[C@@H](O)CNCCNC(=O)N3CCOCC3)cc2)O1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C25H39N3O8.ClH/c1-25(2)35-18-22(36-25)17-34-23(30)8-5-19-3-6-21(7-4-19)33-16-20(29)15-26-9-10-27-24(31)28-11-13-32-14-12-28;/h3-4,6-7,20,22,26,29H,5,8-18H2,1-2H3,(H,27,31);1H/t20-,22+;/m0./s1
Formula Weight 545.2504
Chemical Name or Material Landiolol Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.