Learn More
American Educational Products Landform Demonstration Kit
Demonstrate folding, faulting and a volcanic eruption.
Brand:  American Educational Products 555
Product Code. 13129390
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Includes: Nonflammable volcano, demonstration tray, colored foam, simulated rock formations and a student study guide
Description
Description
  • From an erupting volcano to folding and faulting models
  • Spectacular visual demonstration of the forces that shape the Earth's surface
  • Explains geological terminology such as horst, graben and anticline
  • Nonflammable volcano: 6.7 × 8.66 in. (17 × 22cm)
  • Watertight demonstration tray (L × W × D): 16.9 × 5.12 × 1.97 in. (43 × 13 × 5cm)