missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LANCL2 Antibody (OTI1C5), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72054AF350
This item is not returnable.
View return policy
Description
LANCL2 Monoclonal antibody specifically detects LANCL2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| LANCL2 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| G protein-coupled receptor 69B, GPR69B, LanC (bacterial lantibiotic synthetase component C)-like 2, LanC lantibiotic synthetase component C-like 2 (bacterial), lanC-like protein 2, TASPMGC87139, Testis-specific adriamycin sensitivity protein | |
| Recombinant protein fragment corresponding to amino acids 212-450 of human LANCL2 (NP_061167) produced in E.coli. | |
| 0.1 mL | |
| Signal Transduction | |
| 55915 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI1C5 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction