missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ LANCL1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29508
This item is not returnable.
View return policy
Description
LANCL1 Polyclonal antibody specifically detects LANCL1 in Human,Mouse samples. It is validated for Western Blot
Specifications
| LANCL1 | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| 40 kDa erythrocyte membrane protein, LanC (bacterial lantibiotic synthetase component C)-like 1, LanC (bacterial lantibiotic synthetase component), LanC lantibiotic synthetase component C-like 1 (bacterial), lanC-like protein 1, p40GPR69AG protein-coupled receptor 69A | |
| Between 349 and 399 | |
| 100 μg | |
| Cardiovascular Biology | |
| 10314 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction