missing translation for 'onlineSavingsMsg'
Learn More

Lancer™ LanceRinse Machine Acid Rinse Additive

Product Code. 13132230
Change view
Click to view available options
Volume (Metric):
10 L
Unit Size:
10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
13132230 10 L 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13132230 Supplier Lancer™ Supplier No. 41010014
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Use in both Lancer and other quality washing machines. Lancer™ Rinse Machine Acid Rinse Additive has been specially formulated to give the best possible results.

  • Chemical cleaning compound
  • Health and Safety Data Sheets available to comply with CHP regulations
.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Machine Acid Rinse Additive
For Use With (Application) Removes all alkaline and other deposits during the rinse cycle
For Use With (Equipment) Laboratory glassware washers and hospital instrument washer-disinfectors
Volume (Metric) 10 L
compliance-icons
Product Identifier
  • NLL
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P264-Wash hands thoroughly after handling.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contain: Acetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.