missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Logistik™ Lancer-Style Chemical Splash Laboratory Coats
Provide the answer to full frontal protection with Logistik™ Lancer-Style Chemical Splash Laboratory Coats The wrap-over style Lancer front coat helps repel liquid chemicals and provides the wearer with an excellent level of protection since the use of chemicals in a laboratory can prove a constant hazard to standard style open neck coats.
£74.35 - £76.00
Specifications
Material | Polyester, Carbon |
---|---|
Color | White |
Cuff Style | Stud-fastened |
No. of Pockets | 2 |
Collar Type | Open |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
12834382
|
Logistik™
849A-E2WH-92CM |
Small |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12824382
|
Logistik™
849A-E2WH-100CM |
Medium |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12814382
|
Logistik™
849A-E2WH-108CM |
Large |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Full frontal protection
- Chem-Tech material
- Fluorochemical liquid repellent
Specifications
Polyester, Carbon | |
Stud-fastened | |
Open | |
CE EN 13034 | |
65% Polyester/35% Cotton |
White | |
2 | |
Fluorochemical liquid repellent | |
Designed for Chem-Tech material which helps repel liquid chemicals and provides the wearer with an excellent level of protection |