Learn More
Logistik™ Lancer-Style Chemical Splash Laboratory Coats
Product Code. 12814382
Change view
Click to view available options
Size:
Small
Medium
Large
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
12814382 Large Each
12834382 Small Each
12824382 Medium Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12814382 Supplier Logistik™ Supplier No. 849AE2WH108CM
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Provide the answer to full frontal protection with Logistik™ Lancer-Style Chemical Splash Laboratory Coats The wrap-over style Lancer front coat helps repel liquid chemicals and provides the wearer with an excellent level of protection since the use of chemicals in a laboratory can prove a constant hazard to standard style open neck coats.
  • Full frontal protection
  • Chem-Tech material
  • Fluorochemical liquid repellent
Specifications
Product Type General Purpose Lab Coat
Size Large
Gender Unisex
Material Polyester, Carbon
Color White
Cuff Style Stud-fastened
Disposable Reusable
Sterility Non-sterile
Style Lancer
No. of Pockets 2
Collar Type Open
Finish Fluorochemical Liquid Repellent
Size (Metric) Chest 106.68 to 111.76 cm
Certifications/Compliance CE EN 13034
Length (Metric) 108 cm
Construction 65% Polyester, 35% Cotton
Vis mere Vis mindre
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.