missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LAMTOR5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106356
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total HBXIP.
HBXIP, also named as XIP, belongs to the HBXIP family. It is originally identified by its interaction with the C-terminus of hepatitis B virus (HBV) X protein (HBx). HBXIP is a regulator of centrosome dynamics and cytokinesis. HBXIP is able to up-regulate S100A4 though activating STAT4 and inducing DNA methylation of PTEN. HBXIP is a cellular 18kd protein and cytoplasm stain.
Specifications
| LAMTOR5 | |
| Polyclonal | |
| Unconjugated | |
| LAMTOR5 | |
| 1110003H18Rik; HBV X-interacting protein; HBV X-interacting protein homolog; HBx-interacting protein; HBX-interacting protein homolog; HBXIP; hepatitis B virus x interacting protein; Hepatitis B virus X-interacting protein; hepatitis B virus x-interacting protein (9.6kD); hepatitis B virus X-interacting protein homolog; LAMTOR5; Late endosomal/lysosomal adaptor and MAPK and MTOR activator 5; late endosomal/lysosomal adaptor, MAPK and MTOR activator 5; MGC71071; ragulator complex protein LAMTOR5; XIP | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10542, 295357, 68576 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O43504, Q9D1L9 | |
| LAMTOR5 | |
| A synthesized peptide derived from human LAMTOR5(Accession O43504), corresponding to amino acid residues L5-L55. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction