missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LAMTOR2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552056
This item is not returnable.
View return policy
Description
Immunogen sequence: RPKALTQVLS QANTGGVQST LLLNNEGSLL AYSGYGDTDA RVTAAIASNI WAAYDRNGNQ AFNEDNLKFI LMDCMEGRVA ITRVANLLLC MYAKETVGFG MLKAKAQALV QYLEEP Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
The late endosomal/lysosomal adaptor MAPK and MTOR activator 2 (LAMTOR2) protein belongs to the LAMTOR family of proteins, and together with LAMTOR3 and the MAPK1 and ERK kinase 1 (MEK1) localizes to late endosomes where it is required for the efficient activation of ERK signaling. This complex is involved in the regulation of late endosomal traffic and cellular proliferation and plays a role in cellular host defense against Salmonella infection.
Specifications
| LAMTOR2 | |
| Polyclonal | |
| Unconjugated | |
| LAMTOR2 | |
| 2010111E04Rik; AL022628; ENDAP; Endosomal adaptor protein p14; HSPC003; LAMTOR2; Late endosomal/lysosomal adaptor and MAPK and MTOR activator 2; late endosomal/lysosomal adaptor, MAPK and MTOR activator 2; late endosomal/lysosomal MP1 interacting protein; late endosomal/lysosomal Mp1-interacting protein; MAPBP-interacting protein; Mapbpip; MAPKSP1 adaptor protein; MAPKSP1AP; mitogen activated protein binding protein interacting protein; mitogen-activated protein binding protein interacting protein; mitogen-activated protein-binding protein-interacting protein; mitogen-activated protein-binding protein-interacting protein (MAPBPIP); P14; Rab25; ragulator complex protein LAMTOR2; Ragulator2; RGD1562501; roadblock domain containing 3; roadblock domain-containing protein 3; ROBLD3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 28956 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y2Q5 | |
| LAMTOR2 | |
| Recombinant protein corresponding to Human LAMTOR2. Recombinant protein control fragment (Product #RP-102170). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction