missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
LAMTOR1 Polyclonal specifically detects LAMTOR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | LAMTOR1 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | c11orf59, chromosome 11 open reading frame 59, FLJ20625, LAMTOR1, Late endosomal/lysosomal adaptor and MAPK and MTOR activator 1, late endosomal/lysosomal adaptor, MAPK and MTOR activator 1, Lipid raft adaptor protein p18, p18, p27kip1 releasing factor from RhoA, p27Kip1-releasing factor from RhoA, p27RF-RhoC11orf59, PDRO, PDROPdro, Protein associated with DRMs and endosomes, ragulator complex protein PDRO, Ragulator1, RhoA activator C11orf59 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-161 of human LAMTOR1 (NP_060377.1). |
| Purification Method | Affinity purified |
| Quantity | 100 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?