Learn More
Motic™ Lamphouse HBO 100W
Click to view available options
For Use With (Equipment):
AE30/AE31 Inverted and BA410 Upright Compound Microscopes
Unit Size:
Each
Description
- With collector lens and mercury lamp socket
- For AE30/AE31 inverted and BA410 upright compound microscopes.
- Part of the Complete Epi-Fluorescence Attachment for AE kit (1101000202182).
Specifications
Specifications
| Type | Lamphouse HBO 100W |
| For Use With (Equipment) | AE30/AE31 Inverted and BA410 Upright Compound Microscopes |
| Includes | Collector lens and mercury lamp socket |