Learn More
Lampalizumab Recombinant Human Monoclonal Antibody, Invitrogen™
Human Recombinant Monoclonal Antibody
Brand: Invitrogen MA541971
Additional Details : Weight : 0.01000kg
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Lampalizumab has been used in trials studying the treatment of Geographic Atrophy.Specifications
Lampalizumab | |
Recombinant Monoclonal | |
Unconjugated | |
Anti-fD; AFD; FCFD4514S; RG7417 | |
Human CFD | |
100 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
ELISA | |
0.67 mg/mL | |
PBS with no preservative; pH 7.4 | |
Human | |
Protein L | |
RUO | |
Chemical | |
Antibody | |
IgG1 F(ab) κ |