missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAMPA, 1mg/ml in Acetonitrile, Lipomed
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Description
LAMPA, 1mg/ml in Acetonitrile
Specifications
Specifications
| Chemical Name or Material | Lysergic Acid N-Methyl-N-propylamide |
| CAS | 40158-98-3 |
| Analyte or Component Names | Lysergic Acid N-Methyl-N-propylamide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H25 N3 O |
| Concentration | 1 mg/mL |
| Synonym | (8β)-9,10-Didehydro-N,6-dimethyl-N-propylergoline-8-carboxamide, LAMPA, Lysergic acid methylpropylamide, Lysergic Acid N-Methyl-N-propylamide, N-Methyl-N-propyl-D-lysergamide, N-Methyl-N-propyllysergamide |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | CCCN(C)C(=O)[C@H]1 CN(C)[C@@H]2 Cc3c[nH]c4cccc(C2=C1)c34 |
| IUPAC Name | (6 aR,9 R)-N,7-dimethyl-N-propyl-6,6 A,8,9-tetrahydro-4 H-indolo[4,3-fg]quinoline-9-carboxamide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction