Learn More
Invitrogen™ LAMP2 Polyclonal Antibody

Description
PA1-655 detects lysosome-associated membrane protein-2 (LAMP2) from rat, mouse and human tissues. PA1-655 has been successfully used in Western blot, immunofluorescence, and immunohistochemistry. By Western blot, this antibody detects glycosylated and non-glycosylated LAMP-2, depending on the lysis buffer used. PA1-655 immunizing peptide corresponds to amino acid residues 400-411 from rat LAMP2. This sequence is completely conserved between rat, mouse, and guinea pig LAMP2. PA1-655 immunizing peptide (Cat. # PEP-039) is available for use in neutralization and control experiments.
Specifications
Specifications
| Antigen | LAMP2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA and 0.05% sodium azide |
| Gene | LAMP2 |
| Gene Accession No. | P13473, P17046, P17047 |
| Gene Alias | CD107 antigen-like family member B; CD107b; EGK_20887; Igp96; Lamp II; LAMP2; LAMP-2; LAMP-2 Isoform 2A; LAMP-2 Variant A; Lamp-2a; Lamp-2b; Lamp-2c; LAMPB; LGP110; LGP-110; LGP-96; LGP-B; lysosomal associated membrane protein 2; lysosomal membrane glycoprotein 2; lysosomal membrane glycoprotein type B; lysosomal-associated membrane protein 2; lysosome-associated membrane glycoprotein 2; lysosome-associated membrane protein 2; Mac3; RP23-193O17.2 |
| Gene Symbols | LAMP2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.