missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAMP2 Mouse anti-Human, Clone: 2D3B9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66301-1-IG-150UL
This item is not returnable.
View return policy
Description
Lysosomes are membrane-bound vacuoles which play a critical role in cellular metabolism. These organelles participate in endocytosis, the synthesis and assembly of acid hydrolases, act as sites for digestion of foreign materials and for specialized autolytic cellular processes. Membrane-associated glycoproteins have an important role in normal lysosomal function. Prior to posttranslational modification, Lysosome Associated Membrane Protein 2 (LAMP 2) is an approximately 45 kDa polypeptide. Mature, functional LAMP 2 is extensively glycosylated with a variety of different N-linked and O-linked oligosaccharides with a total molecular weight of approximately 100-110 kDa. LAMP 2 has a structure comprised of a large amino-terminal intra-lysosomal domain, a hydrophobic trans-membrane domain, and a short carboxyl-terminal cytoplasmic tail.Specifications
| LAMP2 | |
| Monoclonal | |
| 0.7 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P13473 | |
| LAMP2 | |
| LAMP2 Fusion Protein Ag0595 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgA |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 2D3B9 | |
| Unconjugated | |
| LAMP2 | |
| CD107b, LAMP 2, LAMP2, LAMPB, LGP110 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 3920 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction