missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LAMP1 Monoclonal Antibody (07), Invitrogen™

Mouse Monoclonal Antibody
£104.00 - £313.00
Specifications
| Antigen | LAMP1 |
|---|---|
| Clone | 07 |
| Concentration | 1 mg/mL |
| Applications | ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30019444
![]() |
Invitrogen™
MA552096 |
100 μL |
£313.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30019438
|
Invitrogen™
MA552097 |
20 μL |
£104.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LAMP1 Monoclonal antibody specifically detects LAMP1 in Human samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), ImmunoprecipitationSpecifications
| LAMP1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 20mM HEPES with 150mM NaCl, 8% sucrose, 0.02% Tween 80 and no preservative; pH 8.0 | |
| 120 kDa lysosomal membrane glycoprotein; AI196048; CD107 antigen-like family member A; CD107a; I79_011073; Lamp I; LAMP1; LAMP-1; LAMPA; LGP120; LGP-120; LGPA; LGP-A; lysosomal associated membrane protein 1; Lysosomal associated membrane protein 1 (120 kDa); lysosomal membrane glycoprotein 1; lysosomal membrane glycoprotein A; lysosomal-associated membrane protein 1; lysosome-associated membrane glycoprotein 1; LYSOSOME-ASSOCIATED MEMBRANE GLYCOPROTEIN 1 PRECURSOR (LAMP-1) (LGP-A) (LGP-120) (CD107A) (P2B); lysosome-associated membrane protein 1; P2B | |
| Recombinant Human LAMP1 protein (Ala 29-Met382). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| LAMP1 |
| 07 | |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P11279 | |
| 3916 | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title