Learn More
Motic™ Lamp Centering Tool, BA410
Product Code. 15485302
Change view
Click to view available options
For Use With (Equipment):
BA410 Microscope
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
15485302 BA410 Microscope Each
1 options
Product Code. 15485302 Supplier Motic™ Supplier No. 1101001701801
Adjust your light source lamp horizontally and vertically with this centering, alignment tool. Motic™ Lamp Centering Tool, AE30, AE31 is part of an EF-UPR epifluorescence attachment kit, but can be purchased separately.
  • Crosshair, patterned centering tool.
  • Facilitates horizontal and vertical lamp centering.
  • Results in evenly distributed and symmetrical beam.
Specifications
Type Lamp Centering Tool
For Use With (Application) EF-UPR epifluorescence
For Use With (Equipment) BA410 Microscope