missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAMP-1/CD107a Antibody (LY1C6), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
£139.00
Specifications
| Antigen | LAMP-1/CD107a |
|---|---|
| Clone | LY1C6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500, Electron Microscopy 1:10-1:500 |
| Applications | Western Blot |
Description
LAMP-1/CD107a Monoclonal specifically detects LAMP-1/CD107a in Human, Mouse, Rat, Hamster samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Electron Microscopy.Specifications
| LAMP-1/CD107a | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Endosome Markers, Mast Cell Markers, Membrane Trafficking and Chaperones | |
| PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide | |
| CD107 antigen-like family member A, CD107a, CD107a antigen, LAMP-1, LAMPA, LGP120, lysosomal-associated membrane protein 1, lysosome-associated membrane glycoprotein 1, Lysosome-associated membrane protein 1 | |
| LAMP1 | |
| IgG1 | |
| Protein G purified | |
| Detects approx 120kDa. |
| LY1C6 | |
| Western Blot 1 μg/mL, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500, Electron Microscopy 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Rat | |
| P14562 | |
| 3916 | |
| Rat liver lysosomal membrane preparations | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title