missing translation for 'onlineSavingsMsg'
Learn More

Lamotrigine, TRC

Product Code. 18093200
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18093200 50 mg 50mg
18080710 100 mg 100mg
18010720 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18093200 Supplier Toronto Research Chemicals Supplier No. TRCL17325050MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84057-84-1
Molecular Formula C9 H7 Cl2 N5
Molecular Weight (g/mol) 256.09
Synonym 1,2,4-Triazine-3,5-diamine, 6-(2,3-dichlorophenyl)- (9CI, ACI), 6-(2,3-Dichlorophenyl)-1,2,4-triazine-3,5-diamine (ACI), 3,5-Diamino-6-(2,3-dichlorophenyl)-1,2,4-triazine, 3,5-Diamino-6-(2,3-dichlorophenyl)-as-triazine, BW 430C, Epilepax, Lamictal, Lamictal XR, Lamidus, Lamitrin, Lamotrigin, Lamotrigine, Lamotrix, Lamtox, LTG
IUPAC Name 6-(2,3-dichlorophenyl)-1,2,4-triazine-3,5-diamine
SMILES Nc1nnc(c(N)n1)c2cccc(Cl)c2Cl

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C9H7Cl2N5/c10-5-3-1-2-4(6(5)11)7-8(12)14-9(13)16-15-7/h1-3H,(H4,12,13,14,16)
Formula Weight 255.0079
Percent Purity >95%
Chemical Name or Material Lamotrigine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.