missing translation for 'onlineSavingsMsg'
Learn More

Lamotrigine, MedChemExpress

Product Code. 16420958
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16420958

Brand: Medchem Express HYB0495

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Lamotrigine (BW430C) is a potent and orally active anticonvulsant or antiepileptic agent. Lamotrigine selectively blocks voltage-gated Na+ channels, stabilizing presynaptic neuronal membranes and inhibiting glutamate release. Lamotrigine can be used for the research of epilepsy, focal seizure, et al.

Lamotrigine (BW430C) is a potent and orally active anticonvulsant or antiepileptic agent. Lamotrigine selectively blocks voltage-gated Na+ channels, stabilizing presynaptic neuronal membranes and inhibiting glutamate release. Lamotrigine can be used for the research of epilepsy, focal seizure, et al.

Specifications

Chemical Name or Material Lamotrigine
Physical Form Solid
CAS 84057-84-1
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C9H7Cl2N5
For Use With (Application) Neuroscience-Neuromodulation
Synonym LTG BW430C
Solubility Information DMSO : 25 mg/mL (97.62 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.39 mM; Need ultrasonic)
SMILES NC1=NC(N)=C(C2=CC=CC(Cl)=C2Cl)N=N1
Molecular Weight (g/mol) 256.09
Formula Weight 256.09
Percent Purity 99.75%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.