missing translation for 'onlineSavingsMsg'
Learn More

Lamotrigine, MedChemExpress

Product Code. 16462199
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16462199 1 g 1g
16420958 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16462199 Supplier Medchem Express Supplier No. HYB0495
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Lamotrigine (BW430C) is a potent and orally active anticonvulsant or antiepileptic agent. Lamotrigine selectively blocks voltage-gated Na+ channels, stabilizing presynaptic neuronal membranes and inhibiting glutamate release. Lamotrigine can be used for the research of epilepsy, focal seizure, et al.

Lamotrigine (BW430C) is a potent and orally active anticonvulsant or antiepileptic agent. Lamotrigine selectively blocks voltage-gated Na+ channels, stabilizing presynaptic neuronal membranes and inhibiting glutamate release. Lamotrigine can be used for the research of epilepsy, focal seizure, et al.

Specifications

Chemical Name or Material Lamotrigine
Physical Form Solid
CAS 84057-84-1
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C9H7Cl2N5
For Use With (Application) Neuroscience-Neuromodulation
Synonym LTG BW430C
Solubility Information DMSO : 25 mg/mL (97.62 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.39 mM; Need ultrasonic)
SMILES NC1=NC(N)=C(C2=CC=CC(Cl)=C2Cl)N=N1
Molecular Weight (g/mol) 256.09
Formula Weight 256.09
Percent Purity 99.75%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.