missing translation for 'onlineSavingsMsg'
Learn More

Lamotrigine, Lipomed

Product Code. 30460013
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30460013 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30460013 Supplier Lipomed Supplier No. LPMLMT1296FB100
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Lamotrigine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lamotrigine
CAS 84057-84-1
Analyte or Component Names Lamotrigine
Shipping Condition Room Temperature
Molecular Formula C9 H7 Cl2 N5
Synonym Lamotrigine, 6-(2,3-Dichlorophenyl)-1,2,4-triazine-3,5-diamine, 3,5-Diamino-6-(2,3-dichlorophenyl)-1,2,4-triazine, BW 430 C, LTG, GI 267119 X, GI 267119 X, GI 267119 X, GI 267119 X, GI 267119 X, GI 267119 X, GI 267119 X, GI 267119 X
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES Nc1nnc(c(N)n1)c2cccc(Cl)c2Cl
IUPAC Name 6-(2,3-dichlorophenyl)-1,2,4-triazine-3,5-diamine
Industry Type Pharmaceutical
Molecular Weight (g/mol) 256.09
Quantity 100 mg
Formula Weight 255.0079 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.