missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Lamotrigine isethionate
GREENER_CHOICE

Product Code. 18772744
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18772744 50 mg 50mg
18772614 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18772744 Supplier Tocris Bioscience™ Supplier No. 2289/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Inhibits glutamate release. Water-soluble salt of lamotrigine (Cat. No. 1611)

Water-soluble salt of lamotrigine (Cat. No. 1611). Displays anticonvulsant effects and inhibits glutamate release, possibly through inhibition of Na+, K+ and Ca2+ currents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 113170-86-8
Molecular Formula C11H13Cl2N5O4S

Specifications

Quantity 50 mg
Formula Weight 382.22
Purity >99%
Molecular Weight (g/mol) 382.216
Chemical Name or Material Lamotrigine isethionate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.