missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Lamotrigine isethionate

Description
Water-soluble salt of lamotrigine (Cat. No. 1611). Displays anticonvulsant effects and inhibits glutamate release, possibly through inhibition of Na+, K+ and Ca2+ currents.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 382.22 |
| Purity | >99% |
| Molecular Weight (g/mol) | 382.216 |
| Chemical Name or Material | Lamotrigine isethionate |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?