missing translation for 'onlineSavingsMsg'
Learn More

Lamotrigine, 98%

Product Code. 15658695 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15658695 5 g 5g
15648695 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15658695 Supplier Thermo Scientific Acros Supplier No. 460850050
  • ONLINE EXCLUSIVE PRICE
    £115.00
    £103.65 / 5g
    Save £11.35
    9% Off
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 84057-84-1 | C9H7Cl2N5 | 256.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84057-84-1
Molecular Formula C9H7Cl2N5
Molecular Weight (g/mol) 256.09
MDL Number MFCD00865333
InChI Key PYZRQGJRPPTADH-UHFFFAOYSA-N
Synonym lamotrigine, 6-2,3-dichlorophenyl-1,2,4-triazine-3,5-diamine, lamictal, lamictal cd, lamotrigina, lamotriginum, lamictal xr, lamotriginum latin, lamotrigina spanish, lamictal odt
PubChem CID 3878
ChEBI CHEBI:6367
IUPAC Name 6-(2,3-dichlorophenyl)-1,2,4-triazine-3,5-diamine
SMILES NC1=NN=C(C(N)=N1)C1=CC=CC(Cl)=C1Cl

Specifications

Melting Point 218.0°C
Loss on Drying 0.5% max. (105°C, 3 h)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in dmf
Formula Weight 256.09
Percent Purity 98%
Chemical Name or Material Lamotrigine
compliance-icons
Product Identifier
  • Lamotrigine, 98%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Supplemental information
  • MIXTURE LIST-Contain: Lamotrigine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.