missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Lamotrigine

Product Code. 18792764
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18792764 50 mg 50mg
18792634 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18792764 Supplier R&D Systems™ Supplier No. 1611/50
  • ONLINE EXCLUSIVE PRICE
    £624.12
    £559.65 / 50mg
    Save £64.47
    10% Off
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Inhibits glutamate release. Anticonvulsant

Anticonvulsant. Inhibits glutamate release, possibly through inhibition of Na+, K+ and Ca2+ currents. Also blocks heterologously expressed and native α4β2 nAChRs with a similar affinity to Na+ channels.

Water-soluble Salt also available.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84057-84-1
Molecular Formula C9H7Cl2N5

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 256.09
Purity >99%
Molecular Weight (g/mol) 256.09
Chemical Name or Material Lamotrigine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.