Learn More
Laminin beta 3 Antibody (CL3353) [mFluor Violet 500 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-46622MFV500
Product Code. 30298681
  • £599.00 / 0.10mL
Description
Laminin beta 3 Monoclonal antibody specifically detects Laminin beta 3 in Human,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Laminin beta 3
Monoclonal
mFluor Violet 500 SE
BM600-125KDA, Epiligrin subunit bata, FLJ99565, Kalinin B1 chain, Kalinin subunit beta, kalinin-140kDa, LAM5, Laminin 5, Laminin B1k chain, laminin S B3 chain, laminin subunit beta-3, laminin, beta 3, laminin, beta 3 (nicein (125kD), kalinin (140kD), BM600 (125kD)), Laminin-5 subunit beta, LAMNB1, Nicein subunit beta, nicein-125kDa
This antibody was developed using a recombinant protein derived from Q13751, with the exact immunogen sequence remaining proprietary.
0.1 mL
Primary
Human, Rat
Purified
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL3353
50mM Sodium Borate
Mouse
Protein A purified
RUO
3914
Store at 4⪚C in the dark.
IgG1