missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Laminin beta-2 Polyclonal Antibody

Laminin beta-2/gamma-1 Polyclonal Antibody
Brand: Invitrogen PA5103211
This item is not returnable.
View return policy
Description
Laminin beta-2/gamma-1 Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| Laminin beta-2 | |
| Polyclonal | |
| Unconjugated | |
| LAMB2 | |
| AW211941; Lamb2; Lamb-2; Laminin B1s chain; laminin chain B3; laminin chain beta 2; Laminin S; laminin subunit beta 2; laminin subunit beta-2; laminin, beta 2; laminin, beta 2 (laminin S); laminin-11 subunit beta; Laminin-14 subunit beta; laminin-15 subunit beta; Laminin-3 subunit beta; laminin-4 subunit beta; Laminin-7 subunit beta; laminin-9 subunit beta; LAMS; NPHS5; npht; SLAM; S-LAM beta; S-laminin subunit beta | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 16779, 25473, 3913 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P15800, P55268, Q61292 | |
| LAMB2 | |
| A synthesized peptide derived from human LAMB2(Accession P55268), corresponding to amino acid residues G72-A122. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction