Learn More
Laminin beta-1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5121243
Description
Positive test controls include: SW480, HepG2, HT-1080, SKOV3, Mouse heart. The target is usually found in the following locations: Secreted, basement membrane, extracellular matrix, extracellular space. Immunogen sequence: EAKLRADEAK QSAEDILLKT NATKEKMDKS NEELRNLIKQ IRNFLTQDSA DLDSIEAVAN EVLKMEMPST PQQLQNLTED IRERVESLSQ VEVILQHSAA DIARAEMLLE EAKRASKSAT DVKVTADMVK EALEEAEKAQ VAAEKAIKQA DEDIQGTQNL LTSIESETAA SEETLFNASQ RISELERNVE ELKRKAAQNS GEAEYIEKVV YTVKQSAEDV KKTLDGELDE KYKKVENLIA KKTEESADAR RKAEMLQNEA KTLLAQANSK LQLLKDLERK YEDNQRYLED KAQELARLEG EVRSLLKDIS QKVAVYSTCL
Laminins are large heterotrimeric, noncollagenous glycoproteins composed of alpha, beta, and gamma chains. They are ubiquitously present in basement membrane (BM) along with entactin/nidogen (EN), collagen type IV (CIV), and large heparan sulfate proteoglycan (HSPG), which interact specifically with each other to form a continuous and regular BM. Alterations of BM integrity, from local discontinuities up to complete loss, are described in many types of human and animal epithelial neoplasms.
Specifications
| Laminin beta-1 | |
| Polyclonal | |
| Unconjugated | |
| LAMB1 | |
| C77966; C80098; C81607; CLM; D130003D08Rik; I79_017505; LAMB1; Lamb-1; LAMB1; basement membrane protein; Lamb1-1; laminin B1; laminin B1 chain; laminin B1 subunit 1; laminin beta 1; laminin beta-1 chain; laminin subunit beta 1; laminin subunit beta-1; laminin, beta 1; laminin-1 subunit beta; Laminin-10 subunit beta; laminin-12 subunit beta; Laminin-2 subunit beta; laminin-6 subunit beta; Laminin-8 subunit beta; LIS5; LOW QUALITY PROTEIN: laminin subunit beta-1; MGC142015 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 16777, 298941, 3912 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunocytochemistry, Western Blot | |
| 0.52 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P02469, P07942 | |
| LAMB1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1467-1786 of human Laminin beta 1 (NP_002282.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.