missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Laminin beta-1 Monoclonal Antibody (LT3), Invitrogen™
Description
MA5-14657 targets Laminin B1/Beta1 in IF, IHC (P, F), IP, and WB applications and shows reactivity with Hamster, Human, mouse, and Porcine samples. The MA5-14657 immunogen is mouse EHS laminin preparation.
Laminin beta-1 Monoclonal antibody specifically detects Laminin beta-1 in Hamster, Human, Mouse, Porcine, Rat samples. It is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.
Specifications
Specifications
| Antigen | Laminin beta-1 |
| Applications | Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | LT3 |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 |
| Gene | LAMB1 |
| Gene Accession No. | P02469, P07942 |
| Gene Alias | C77966; C80098; C81607; CLM; D130003D08Rik; I79_017505; LAMB1; Lamb-1; LAMB1; basement membrane protein; Lamb1-1; laminin B1; laminin B1 chain; laminin B1 subunit 1; laminin beta 1; laminin beta-1 chain; laminin subunit beta 1; laminin subunit beta-1; laminin, beta 1; laminin-1 subunit beta; Laminin-10 subunit beta; laminin-12 subunit beta; Laminin-2 subunit beta; laminin-6 subunit beta; Laminin-8 subunit beta; LIS5; LOW QUALITY PROTEIN: laminin subunit beta-1; MGC142015 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?