missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Laminin beta 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-32043
This item is not returnable.
View return policy
Description
Laminin beta 1 Polyclonal specifically detects Laminin beta 1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Laminin beta 1 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0) and 40% Glycerol with 0.01% Thimerosal | |
| LAMB1 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human Laminin beta 1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 3912 | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| CLM, cutis laxa with marfanoid phenotype, Laminin B1 chain, laminin subunit beta-1, laminin, beta 1, Laminin-1 subunit beta, Laminin-10 subunit beta, Laminin-12 subunit beta, Laminin-2 subunit beta, Laminin-6 subunit beta, Laminin-8 subunit beta, MGC142015 | |
| Rabbit | |
| 198 kDa | |
| 100 μL | |
| Primary | |
| Tested in human. Predicted cross-reactivity based on sequence identity: Bovine (89%). Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction