missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Laminin beta 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35336-20ul
This item is not returnable.
View return policy
Description
Laminin beta 1 Polyclonal antibody specifically detects Laminin beta 1 in Human samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| Laminin beta 1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| CLM, cutis laxa with marfanoid phenotype, Laminin B1 chain, laminin subunit beta-1, laminin, beta 1, Laminin-1 subunit beta, Laminin-10 subunit beta, Laminin-12 subunit beta, Laminin-2 subunit beta, Laminin-6 subunit beta, Laminin-8 subunit beta, MGC142015 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1467-1786 of human Laminin beta 1 (NP_002282.2).,, Sequence:, EAKLRADEAKQSAEDILLKTNATKEKMDKSNEELRNLIKQIRNFLTQDSADLDSIEAVANEVLKMEMPSTPQQLQNLTEDIRERVESLSQVEVILQHSAADIARAEMLLEEAKRASKSATDVKVTADMVKEALEEAEKAQVAAEKAIKQADEDIQGTQNLLTSIESETAASEETLFNASQRISELERNVEELKRKAAQNSGEAEYIEKVVYTVKQSAEDVKKTLDGELDEKYKKVENLIAKKTEESADARRKAEMLQNEAKTLLAQANSKLQLLKDLERKYEDNQRYLEDKAQELARLEGEVRSLLKDISQKVAVYSTCL | |
| 20 μL | |
| Angiogenesis, Apoptosis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Neuroscience, Signal Transduction, Tumor Suppressors | |
| 3912 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction