missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Laminin alpha-5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100745
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Antibody detects endogenous levels of total LAMA5.
Components of the extracellular matrix exert myriad effects on tissues throughout the body. In particular, the laminins, a family of heterotrimeric extracellular glycoproteins, affect tissue development and integrity in such diverse organs as the kidney, lung, skin, and nervous system. It is thought that laminins mediate the attachment, migration, and organization of cells into tissues during embryonic development by interacting with other extracellular matrix components. Laminins function as heterotrimeric complexes of alpha, beta, and gamma chains, with each chain type representing a different subfamily of proteins. The protein encoded by this gene belongs to the alpha subfamily of laminin chains and is a major component of basement membranes. Two transcript variants encoding different isoforms have been found for this gene, but the full-length nature of one of them has not been determined.
Specifica
| Laminin alpha-5 | |
| Polyclonal | |
| Unconjugated | |
| LAMA5 | |
| [a]5; AA408760; AA408762; AI853660; KIAA0533; KIAA1907; LAMA5; laminin alpha-5 chain; laminin subunit alpha 5; laminin subunit alpha-5; laminin, alpha 5; laminin-10 subunit alpha; Laminin-11 subunit alpha; laminin-15 subunit alpha; laminin-511; mKIAA0533; RP11-157P1.6 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 16776, 3911 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O15230, Q61001 | |
| LAMA5 | |
| A synthesized peptide derived from human LAMA5(Accession O15230), corresponding to amino acid residues V512-C562. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto