Learn More
Thermo Scientific Chemicals Laminin (925-933)
Description
Derived from residues 925-933 of human laminin B1, this peptide binds the laminin receptor, Inhibits experimental metastasis. find useful for product applications. And it is also used in Western Blot, Blocking Peptides, Dot Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Flow Cytomety, Cell Culture.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Purification Method | HPLC |
| Content And Storage | -30°C to -10°C |
| Format | Solid |
| For Use With (Application) | Derived from residues 925-933 of human laminin B1, this peptide binds the laminin receptor |
| Molecular Weight (g/mol) | 967.05 |
| Product Type | Laminin (925-933) |
| Quantity | 0.5 mg |
| Regulatory Status | RUO - research use only |
| Sequence | Cys-Asp-Pro-Gly-Tyr-Ile-Gly-Ser-Arg |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.