missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Laminin-213 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114757
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total LAMC3.
Laminin is a non-collagenous protein of basement membranes. By rotary shadowing it has a cross-like shape, consisting of one long arm, (400 kD) and three short arms (200 kD). Antibodies to laminin localize exclusively in basement membranes. Laminin is found throughout the basement membrane, but the reaction with the antibodies to laminin is more intense in the lamina lucida, the part of the basement membrane adjacent to the cell membrane. Laminin binds to type IV collagen, a component exclusively localized in the basement membrane. Laminin has been shown to play a role in cell adhesion and attachment in vivo and in vitro.
Specifications
| Laminin-213 | |
| Polyclonal | |
| Unconjugated | |
| LAMC3 | |
| AI562206; AW240805; Lamc3; laminin gamma 3; Laminin gamma3; laminin subunit gamma 3; laminin subunit gamma-3; laminin, gamma 3; laminin-12 subunit gamma; Laminin-14 subunit gamma; laminin-15 subunit gamma; OCCM | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10319 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q9Y6N6 | |
| LAMC3 | |
| A synthesized peptide derived from human LAMC3(Accession Q9Y6N6), corresponding to amino acid residues V497-L547. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction