missing translation for 'onlineSavingsMsg'
Learn More

Laminin-1 Antibody (AL-2) [HRP], Novus Biologicals™

Product Code. 30734236 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30734236 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734236 Supplier Novus Biologicals Supplier No. FAB2549H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Laminin-1 Monoclonal antibody specifically detects Laminin-1 in Mouse samples. It is validated for Immunohistochemistry,Western Blot,Adhesion Blockade
TRUSTED_SUSTAINABILITY

Specifications

Antigen Laminin-1
Applications Immunohistochemistry, Western Blot, Adhesion Blockade
Classification Monoclonal
Clone AL-2
Conjugate HRP
Formulation PBS
Gene Alias LAMA, LAMA1, laminin subunit alpha 1, PTBHS, S-LAM-alpha
Host Species Rat
Immunogen Purified fragment of chymotrypsin-digested mouse EHS tumor-derived Laminin-1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284217
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.