missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Laminated Polyester Warning Signs

Product Code. 10376094
Change view
Click to view available options
Width (Metric):
50 mm
100 mm
Displayed Text:
Corrosive Batteries Warning Pictogram
Radioactive substances 7C III
Warning of Corrosive Substances
Warning of Hazardous Substances
Unit Size:
1 roll
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Displayed Text unitSize
10376094 100 mm Corrosive Batteries Warning Pictogram Each
15138743 50 mm Radioactive substances 7C III 1 roll
10775315 100 mm Warning of Corrosive Substances Each
10403194 100 mm Warning of Hazardous Substances Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10376094 Supplier Brady™ Supplier No. 250231
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Certifications/Compliance ISO 7010, RoHS compliant using EU Directive 2002/95/EC, ROHS Environmental Compliance
Alternate Part Number PIC 310-TRI 100-B7541
Material Polyester
Quantity 1 Ea.
Adhesive Type Permanent Acrylic
Description Laminated Polyester
For Use With (Application) Pipe markers and safety signs
Height (Metric) 100 mm
Thickness 0.024 mm
Width (Metric) 100 mm
Temperature Range (Metric) -40°C to 110°C
Shape Triangular
Displayed Text Corrosive Batteries Warning Pictogram
Product Type Warning Sign
Color Black on Yellow
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.