missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Laminated Polyester: ISO Safety Sign - Connect an earth terminal to the ground

Sign compliant to ISO 7010.

Brand:  Brady™ 818567

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15694457

  • £5.00 / Pack of 2
Estimated Shipment: 15-03-2024
to see stock.
Add to basket

Description

Description

  • Laminated polyester
  • For indoor and outdoor use
Specifications

Specifications

ISO 7010
Polyester
Permanent Acrylic
Indoor and Outdoor
-40°C to 110°C
Global
Gloss
2/Pk.
50 mm
0.099 mm
Round
White on Blue
Recommendations

Recommendations

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers