missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Laminated Polyester GHS Symbol Signs

Product Code. 15694207
Click to view available options
Width (Metric):
40 mm
50 mm
70 mm
100 mm
Displayed Text:
Acute Toxicity
Compressed Gas
Corrosive
Explosive
Flammable
Hazardous to Aquatic Environment
Health Hazard
Oxidizing
Respiratory Hazard
Style:
Card Mounted
Continuous Roll, 250 Symbols (not labels)/Roll
Individual Markers on a Roll
Quantity:
1 Ea.
100/Pk.
20/Pk.
250/Pk.
6/Pk.
Unit Size:
1 roll
Pack of 20
Pack of 250
Pack of 4
Pack of 6
This item is not returnable. View return policy

Product Code. 15694207

Brand: Brady™ 811691

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Help ensure safety at work and maximize productivity with these GHS warning labels. Brady™ Laminated Polyester GHS Symbol Signs are available in a variety of sizes, with and without text. Standardized images reduce the risk of misunderstandings.

  • Self-adhesive diamond shape labels
  • Pre-printed with GHS hazard symbols
  • Laminated polyester, gloss finish black and red on white

Specifications

Certifications/Compliance ISO 7010, RoHS compliant using EU Directive 2002/95/EC
Alternate Part Number PIC 1803-50*50-B7541-RLL-C
Finish Gloss
Material Polyester
Quantity 250/Pk.
Style Continuous Roll, 250 Symbols (not labels)/Roll
Adhesive Type Permanent Acrylic, Pressure-sensitive
Description Laminated Polyester
For Use With (Application) Pipe markers and safety signs
Height (Metric) 50 mm
Thickness 0.024 mm
Width (Metric) 50 mm
Temperature Range (Metric) -40°C to 110°C
Shape Diamond
Displayed Text Oxidizing
Product Type GHS Symbol Sign
Color Black on White
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.